Merck
  • Neural differentiation from human embryonic stem cells in a defined adherent culture condition.
Neural differentiation from human embryonic stem cells in a defined adherent culture condition.
The International journal of developmental biology (2007-07-10)
Hossein Baharvand, Narges-Zare Mehrjardi, Maryam Hatami, Sahar Kiani, Mahendra Rao, Mahdi-Montazer Haghighi
ABSTRACT
Understanding how to direct human embryonic stem cells (hESCs) toward a specific lineage pathway and generate appropriate cell types robustly is very important, not only for the study of developmental biology but also for potentially using these cells to treat human diseases. In this study, hESCs were differentiated to the neural lineage in defined adherent culture by retinoic acid and basic fibroblast growth factor. Our protocol seems to recapitulate the early steps of nervous system development in vivo in that undifferentiated hESCs organized into rosettes and then neural tube-like structures are formed. Differentiating cells expressed neuroectodermal and mature neuron markers during neural plate and tube formation and maturation, as shown by reverse transcriptase-PCR. More than 90% of differentiated cells expressed additional neuron-specific antigens (i.e., tubulin-III, MAP-2, synaptophysin and neurofilament protein). Ultrastructural analysis of differentiating neural tube-like structures in three dimensional collagen scaffolds showed an ependymal-like layer and neural structure with typical synapses. These results provide a simple and relatively defined system for differentiation of hESCs to neural lineages, particularly neurons with typical cellular, molecular and ultrastuctureal markers. The culture of neural precursor cells in a collagen scaffold may provide a new approach for the repair of spinal cord injury.
Sigma-Aldrich
Monoclonal Anti-β-Tubulin III antibody produced in mouse, clone SDL.3D10, ascites fluid
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
Anti-Nestin Antibody, clone 10C2, clone 10C2, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-Neurofilament 200 antibody produced in mouse, clone NE14, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Synaptophysin antibody produced in mouse, clone SVP-38, ascites fluid
Sigma-Aldrich
Cresyl Violet acetate, certified by the BSC
Sigma-Aldrich
Anti-MAP2 (2a+2b) antibody, Mouse monoclonal, clone AP-20, ascites fluid
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, (H+L) FITC conjugate, 1.15 mg/mL, Chemicon®
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder