Merck
  • miR-92a inhibits peritoneal dissemination of ovarian cancer cells by inhibiting integrin α5 expression.
miR-92a inhibits peritoneal dissemination of ovarian cancer cells by inhibiting integrin α5 expression.
The American journal of pathology (2013-03-19)
Chifumi Ohyagi-Hara, Kenjiro Sawada, Shoji Kamiura, Yasuhiko Tomita, Aki Isobe, Kae Hashimoto, Yasuto Kinose, Seiji Mabuchi, Takeshi Hisamatsu, Toshifumi Takahashi, Keiichi Kumasawa, Shigenori Nagata, Ken-Ichirou Morishige, Ernst Lengyel, Hirohisa Kurachi, Tadashi Kimura
ABSTRACT
Ovarian cancer is characterized by widespread peritoneal dissemination and ascites and has a cure rate of only 30%. As has been previously reported, integrin α5 plays a key role in the peritoneal dissemination of ovarian cancer. Our aim was to identify a new miRNA that regulates integrin α5 expression and analyze the therapeutic potential of targeting this miRNA. By using an IHC analysis, we proved that high integrin α5 expression correlates with a poor prognosis in Japanese patients with International Federation of Gynecology and Obstetrics stage III ovarian cancer. Based on an miRNA algorithm search, we identified hsa-mir-92a (miR-92a) as a candidate. The level of miR-92a expression was significantly inversely correlated with ITGA5 expression in various cancer cells. Transfection of precursor miR-92a reduced integrin α5 expression in ovarian cancer cells, which was accompanied by the inhibition of cancer cell adhesion, invasion, and proliferation. miR-92a overexpression reduced the luciferase activity of the ITGA5 3'-untranslated region, suggesting that ITGA5 mRNA is a direct target of miR-92a. In in vivo ovarian cancer xenografts, the enforced expression of miR-92a in HeyA-8 cells suppressed peritoneal dissemination. Although we still have a long way to go before an effective and nontoxic miRNA-based cancer therapy can be introduced into the clinic, the inhibition of integrin α5 expression by targeting miR-92a needs to be explored further for future applications in ovarian cancer treatment.