Merck
  • m-Bop, a repressor protein essential for cardiogenesis, interacts with skNAC, a heart- and muscle-specific transcription factor.
m-Bop, a repressor protein essential for cardiogenesis, interacts with skNAC, a heart- and muscle-specific transcription factor.
The Journal of biological chemistry (2002-05-16)
Robert J Sims, Elizabeth K Weihe, Li Zhu, Sean O'Malley, June V Harriss, Paul D Gottlieb
ABSTRACT
The m-Bop protein encoded by the mouse Bop gene is strongly expressed in heart and skeletal muscle, and recent studies with Bop knockout mice have demonstrated that m-Bop is essential for cardiogenesis in vivo and can act as a HDAC-dependent repressor in vitro. In the present studies, m-Bop was observed to interact with skNAC, a reported transcriptional activator specific to heart and skeletal muscle. The amino-terminal S region of the split S-ET domain of m-Bop as well as the MYND domain were required for interaction with skNAC in both the two-hybrid system and in coimmunoprecipitation experiments from cultured mammalian cells. As shown previously for interaction of the MYND domain-containing transcriptional corepressor, BS69, with several viral and cellular oncoproteins, a PXLXP motif in skNAC was required for interaction with m-Bop. Similar kinetics of induction and localization of m-Bop and skNAC during the induction of myogenesis in cultured C2C12 cells suggests a possible associated role for these proteins during this process.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M5 antibody produced in mouse, clone M5, purified immunoglobulin, buffered aqueous solution