Merck
  • Synaptic molecular imaging in spared and deprived columns of mouse barrel cortex with array tomography.
Synaptic molecular imaging in spared and deprived columns of mouse barrel cortex with array tomography.
Scientific data (2014-01-01)
Nicholas C Weiler, Forrest Collman, Joshua T Vogelstein, Randal Burns, Stephen J Smith
ABSTRACT
A major question in neuroscience is how diverse subsets of synaptic connections in neural circuits are affected by experience dependent plasticity to form the basis for behavioral learning and memory. Differences in protein expression patterns at individual synapses could constitute a key to understanding both synaptic diversity and the effects of plasticity at different synapse populations. Our approach to this question leverages the immunohistochemical multiplexing capability of array tomography (ATomo) and the columnar organization of mouse barrel cortex to create a dataset comprising high resolution volumetric images of spared and deprived cortical whisker barrels stained for over a dozen synaptic molecules each. These dataset has been made available through the Open Connectome Project for interactive online viewing, and may also be downloaded for offline analysis using web, Matlab, and other interfaces.
Sigma-Aldrich
Anti-Glutamate Receptor 2 Antibody, extracellular, clone 6C4, clone 6C4, Chemicon®, from mouse
Sigma-Aldrich
Anti-γ-Actin antibody, Mouse monoclonal, clone 2-2.1.14.17, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Glutamate receptor 1 Antibody, from rabbit, purified by affinity chromatography