Merck
  • Mutations in MMP9 and MMP13 determine the mode of inheritance and the clinical spectrum of metaphyseal anadysplasia.
Mutations in MMP9 and MMP13 determine the mode of inheritance and the clinical spectrum of metaphyseal anadysplasia.
American journal of human genetics (2009-07-21)
Ekkehart Lausch, Romy Keppler, Katja Hilbert, Valerie Cormier-Daire, Sarah Nikkel, Gen Nishimura, Sheila Unger, Jürgen Spranger, Andrea Superti-Furga, Bernhard Zabel
ABSTRACT
The matrix metalloproteinases MMP9 and MMP13 catalyze the degradation of extracellular matrix (ECM) components in the growth plate and at the same time cleave and release biologically active molecules stored in the ECM, such as VEGFA. In mice, ablation of Mmp9, Mmp13, or both Mmp9 and Mmp13 causes severe distortion of the metaphyseal growth plate. We report that mutations in either MMP9 or MMP13 are responsible for the human disease metaphyseal anadysplasia (MAD), a heterogeneous group of disorders for which a milder recessive variant and a more severe dominant variant are known. We found that recessive MAD is caused by homozygous loss of function of either MMP9 or MMP13, whereas dominant MAD is associated with missense mutations in the prodomain of MMP13 that determine autoactivation of MMP13 and intracellular degradation of both MMP13 and MMP9, resulting in a double enzymatic deficiency.
Sigma-Aldrich
Anti-V5 antibody, Mouse monoclonal, clone V5-10, purified from hybridoma cell culture
Sigma-Aldrich
Anti-MMP-13 Antibody, clone VIIIA2, clone VIIIA2, Chemicon®, from mouse
Roche
Anti-His6, from mouse IgG1