Merck
  • A SPOPL/Cullin-3 ubiquitin ligase complex regulates endocytic trafficking by targeting EPS15 at endosomes.
A SPOPL/Cullin-3 ubiquitin ligase complex regulates endocytic trafficking by targeting EPS15 at endosomes.
eLife (2016-03-24)
Michaela Gschweitl, Anna Ulbricht, Christopher A Barnes, Radoslav I Enchev, Ingrid Stoffel-Studer, Nathalie Meyer-Schaller, Jatta Huotari, Yohei Yamauchi, Urs F Greber, Ari Helenius, Matthias Peter
ABSTRACT
Cullin-3 (CUL3)-based ubiquitin ligases regulate endosome maturation and trafficking of endocytic cargo to lysosomes in mammalian cells. Here, we report that these functions depend on SPOPL, a substrate-specific CUL3 adaptor. We find that SPOPL associates with endosomes and is required for both the formation of multivesicular bodies (MVBs) and the endocytic host cell entry of influenza A virus. In SPOPL-depleted cells, endosomes are enlarged and fail to acquire intraluminal vesicles (ILVs). We identify a critical substrate ubiquitinated by CUL3-SPOPL as EPS15, an endocytic adaptor that also associates with the ESCRT-0 complex members HRS and STAM on endosomes. Indeed, EPS15 is ubiquitinated in a SPOPL-dependent manner, and accumulates with HRS in cells lacking SPOPL. Together, our data indicates that a CUL3-SPOPL E3 ubiquitin ligase complex regulates endocytic trafficking and MVB formation by ubiquitinating and degrading EPS15 at endosomes, thereby influencing influenza A virus infection as well as degradation of EGFR and other EPS15 targets.
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Anti-Rab11 antibody produced in rabbit, 1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Actin Antibody, clone C4, clone C4, Chemicon®, from mouse
Sigma-Aldrich
Anti-EGFR Antibody, Upstate®, from rabbit
Sigma-Aldrich
Monoclonal Anti-HGS antibody produced in mouse, clone 6D11, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP), meets EP testing specifications