Merck
  • HIV Protease Inhibitors Alter Amyloid Precursor Protein Processing via β-Site Amyloid Precursor Protein Cleaving Enzyme-1 Translational Up-Regulation.
HIV Protease Inhibitors Alter Amyloid Precursor Protein Processing via β-Site Amyloid Precursor Protein Cleaving Enzyme-1 Translational Up-Regulation.
The American journal of pathology (2016-12-21)
Patrick J Gannon, Cagla Akay-Espinoza, Alan C Yee, Lisa A Briand, Michelle A Erickson, Benjamin B Gelman, Yan Gao, Norman J Haughey, M Christine Zink, Janice E Clements, Nicholas S Kim, Gabriel Van De Walle, Brigid K Jensen, Robert Vassar, R Christopher Pierce, Alexander J Gill, Dennis L Kolson, J Alan Diehl, Joseph L Mankowski, Kelly L Jordan-Sciutto
ABSTRACT
Mounting evidence implicates antiretroviral (ARV) drugs as potential contributors to the persistence and evolution of clinical and pathological presentation of HIV-associated neurocognitive disorders in the post-ARV era. Based on their ability to induce endoplasmic reticulum (ER) stress in various cell types, we hypothesized that ARV-mediated ER stress in the central nervous system resulted in chronic dysregulation of the unfolded protein response and altered amyloid precursor protein (APP) processing. We used in vitro and in vivo models to show that HIV protease inhibitor (PI) class ARVs induced neuronal damage and ER stress, leading to PKR-like ER kinase-dependent phosphorylation of the eukaryotic translation initiation factor 2α and enhanced translation of β-site APP cleaving enzyme-1 (BACE1). In addition, PIs induced β-amyloid production, indicative of increased BACE1-mediated APP processing, in rodent neuroglial cultures and human APP-expressing Chinese hamster ovary cells. Inhibition of BACE1 activity protected against neuronal damage. Finally, ARVs administered to mice and SIV-infected macaques resulted in neuronal damage and BACE1 up-regulation in the central nervous system. These findings implicate a subset of PIs as potential mediators of neurodegeneration in HIV-associated neurocognitive disorders.
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Millipore
Ticarcillin Supplement, suitable for microbiology
Sigma-Aldrich
Lactacystin, ≥90% (HPLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Anisomycin from Streptomyces griseolus, ≥98% (HPLC), solid
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
β-Secretase Inhibitor IV, β-Secretase Inhibitor IV, CAS 797035-11-1, is a cell-permeable inhibitor that binds to BACE-1 active site and blocks its proteolytic activity (IC50 = 15 nM for human BACE-1).