Merck
  • Angiotensin converting enzyme immobilized on magnetic beads as a tool for ligand fishing.
Angiotensin converting enzyme immobilized on magnetic beads as a tool for ligand fishing.
Journal of pharmaceutical and biomedical analysis (2016-10-12)
Fernando G de Almeida, Kenia L Vanzolini, Quezia B Cass
ABSTRACT
Angiotensin converting enzyme (ACE) presents an important role in blood pressure regulation, since that converts angiotensin I to the vasoconstrictor angiotensin II. Some commercially available ACE inhibitors are captopril, lisinopril and enalapril; due to their side effects, naturally occurring inhibitors have been prospected. In order to endorse this research field we have developed a new tool for ACE ligand screening. To this end, ACE was extracted from bovine lung, purified and chemically immobilized in modified ferrite magnetic beads (ACE-MBs). The ACE-MBs have shown a Michaelian kinetic behavior towards hippuryl-histidyl-leucine. Moreover, as proof of concept, the ACE-MBs was inhibited by lisinopril with a half maximal inhibitory concentration (IC50) of 10nM. At the fishing assay, ACE-MBs were able not only to fish out the reference inhibitor, but also one peptide from a pool of tryptic digested BSA. In conclusion, ACE-MBs emerge as new straightforward tool for ACE kinetics determination, inhibition and binder screening.
Sigma-Aldrich
N-Hexanoyl-L-homoserine lactone, ≥96% (HPLC)
Sigma-Aldrich
Triton X-100, laboratory grade