Merck
Phospho-ubiquitin-PARK2 complex as a marker for mitophagy defects.
Autophagy (2016-11-16)
Sylvie Callegari, Silke Oeljeklaus, Bettina Warscheid, Sven Dennerlein, Michael Thumm, Peter Rehling, Jan Dudek
ABSTRACT
The E3 ubiquitin ligase PARK2 and the mitochondrial protein kinase PINK1 are required for the initiation of mitochondrial damage-induced mitophagy. Together, PARK2 and PINK1 generate a phospho-ubiquitin signal on outer mitochondrial membrane proteins that triggers recruitment of the autophagy machinery. This paper describes the detection of a defined 500-kDa phospho-ubiquitin-rich PARK2 complex that accumulates on mitochondria upon treatment with the membrane uncoupler CCCP. Formation of this complex is dependent on the presence of PINK1 and is absent in mutant forms of PARK2, whereby mitophagy is also arrested. These results signify a functional signaling complex that is essential for the progression of mitophagy. The visualization of the PARK2 signaling complex represents a novel marker for this critical step in mitophagy and can be used to monitor mitophagy progression in PARK2 mutants and to uncover additional upstream factors required for PARK2-mediated mitophagy signaling.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-AUF1 Antibody, Upstate®, from rabbit
Pricing and availability is not currently available.
Sigma-Aldrich
GeneJuice® Transfection Reagent, Non-lipid based chemical transfection reagent optimized for maximum transfection efficiency, ease-of-use, and minimal cytotoxicity on a wide variety of mammalian cells.
Pricing and availability is not currently available.
Sigma-Aldrich
Fluoroshield with DAPI, liquid
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human PINK1, PINK1-AS
Pricing and availability is not currently available.
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Pricing and availability is not currently available.
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Parkin antibody, Mouse monoclonal, clone PRK8, purified from hybridoma cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-ACTB, affinity isolated antibody
Pricing and availability is not currently available.