Merck
  • Anti-apoptotic quinolinate phosphoribosyltransferase (QPRT) is a target gene of Wilms' tumor gene 1 (WT1) protein in leukemic cells.
Anti-apoptotic quinolinate phosphoribosyltransferase (QPRT) is a target gene of Wilms' tumor gene 1 (WT1) protein in leukemic cells.
Biochemical and biophysical research communications (2016-11-28)
Tove Ullmark, Giorgia Montano, Linnea Järvstråt, Helena Jernmark Nilsson, Erik Håkansson, Kristina Drott, Björn Nilsson, Karina Vidovic, Urban Gullberg
ABSTRACT
Wilms' tumor gene 1 (WT1) is a zinc finger transcription factor that has been implicated as an oncogene in leukemia and several other malignancies. When investigating possible gene expression network partners of WT1 in a large acute myeloid leukemia (AML) patient cohort, one of the genes with the highest correlation to WT1 was quinolinate phosphoribosyltransferase (QPRT), a key enzyme in the de novo nicotinamide adenine dinucleotide (NAD+) synthesis pathway. To investigate the possible relationship between WT1 and QPRT, we overexpressed WT1 in hematopoietic progenitor cells and cell lines, resulting in an increase of QPRT expression. WT1 knock-down gave a corresponding decrease in QPRT gene and protein expression. Chromatin-immunoprecipitation revealed WT1 binding to a conserved site in the first intron of the QPRT gene. Upon overexpression in leukemic K562 cells, QPRT conferred partial resistance to the anti-leukemic drug imatinib, indicating possible anti-apoptotic functions, consistent with previous reports on glioma cells. Interestingly, the rescue effect of QPRT overexpression was not correlated to increased NAD + levels, suggesting NAD + independent mechanisms. We conclude that QPRT, encoding a protein with anti-apoptotic properties, is a novel and direct target gene of WT1 in leukemic cells.
Sigma-Aldrich
MISSION® esiRNA, targeting human WT1
Pricing and availability is not currently available.
Sigma-Aldrich
Influenza Hemagglutinin (HA) Peptide, ≥97% (HPLC)
Pricing and availability is not currently available.