Merck
  • The localization of the human myeloperoxidase gene is in close proximity to the translocation breakpoint in acute promyelocytic leukemia.
The localization of the human myeloperoxidase gene is in close proximity to the translocation breakpoint in acute promyelocytic leukemia.
Leukemia (1987)
Chang KS