Merck
  • Experimental Confirmation of Isotope Fractionation in Thiomolybdates Using Ion Chromatographic Separation and Detection by Multicollector ICPMS.
Experimental Confirmation of Isotope Fractionation in Thiomolybdates Using Ion Chromatographic Separation and Detection by Multicollector ICPMS.
Analytical chemistry (2017-02-15)
Carolin F Kerl, Regina Lohmayer, Elvira Bura-Nakić, Derek Vance, Britta Planer-Friedrich
ABSTRACT
Molybdenum
Sigma-Aldrich
Ammonium tetrathiomolybdate, 99.97% trace metals basis
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)