Merck
  • Benzimidazole acrylonitriles as multifunctional push-pull chromophores: Spectral characterisation, protonation equilibria and nanoaggregation in aqueous solutions.
Benzimidazole acrylonitriles as multifunctional push-pull chromophores: Spectral characterisation, protonation equilibria and nanoaggregation in aqueous solutions.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2017-02-16)
Ema Horak, Robert Vianello, Marijana Hranjec, Svjetlana Krištafor, Grace Karminski Zamola, Ivana Murković Steinberg
ABSTRACT
Heterocyclic donor-π-acceptor molecular systems based on an N,N-dimethylamino phenylacrylonitrile benzimidazole skeleton have been characterised and are proposed for potential use in sensing applications. The benzimidazole moiety introduces a broad spectrum of useful multifunctional properties to the system including electron accepting ability, pH sensitivity and compatibility with biomolecules. The photophysical characterisation of the prototropic forms of these chromophores has been carried out in both solution and on immobilisation in polymer films. The experimental results are further supported by computational determination of pK
Sigma-Aldrich
Dioctyl sebacate, AldrichCPR
Supelco
Potassium tetrakis(4-chlorophenyl)borate, Selectophore, ≥98.0%