Merck
  • Multiresponse optimization of a UPLC method for the simultaneous determination of tryptophan and 15 tryptophan-derived compounds using a Box-Behnken design with a desirability function.
Multiresponse optimization of a UPLC method for the simultaneous determination of tryptophan and 15 tryptophan-derived compounds using a Box-Behnken design with a desirability function.
Food chemistry (2017-02-15)
Widiastuti Setyaningsih, Irfan E Saputro, Ceferino A Carrera, Miguel Palma, Carmelo G Barroso
ABSTRACT
A Box-Behnken design was used in conjunction with multiresponse optimization based on the desirability function to carry out the simultaneous separation of tryptophan and 15 derivatives by Ultra Performance Liquid Chromatography. The gradient composition of the mobile phase and the flow rate were optimized with respect to the resolution of severely overlapping chromatographic peaks and the total run time. Two different stationary phases were evaluated (hybrid silica and a solid-core-based C
Sigma-Aldrich
Indole-3-butyric acid potassium salt, suitable for plant cell culture, BioReagent
Supelco
Serotonin, analytical standard
Sigma-Aldrich
1-Methyl-L-tryptophan, 95%