Merck
  • Octopamine and tyramine regulate the activity of reproductive visceral muscles in the adult female blood-feeding bug,
Octopamine and tyramine regulate the activity of reproductive visceral muscles in the adult female blood-feeding bug,
The Journal of experimental biology (2017-02-27)
Sam Hana, Angela B Lange
ABSTRACT
The role of octopamine and tyramine in regulating spontaneous contractions of reproductive tissues was examined in the female
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder