Merck
  • Novel Regulatory Mechanisms for the SoxC Transcriptional Network Required for Visual Pathway Development.
Novel Regulatory Mechanisms for the SoxC Transcriptional Network Required for Visual Pathway Development.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2017-04-16)
Kun-Che Chang, Jonathan Hertz, Xiong Zhang, Xiao-Lu Jin, Peter Shaw, Brooke A Derosa, Janet Y Li, Praseeda Venugopalan, Daniel A Valenzuela, Roshni D Patel, Kristina R Russano, Shomoukh A Alshamekh, Catalina Sun, Kevin Tenerelli, Chenyi Li, Dmitri Velmeshev, Yuyan Cheng, Timothy M Boyce, Alexandra Dreyfuss, Mohammed S Uddin, Kenneth J Muller, Derek M Dykxhoorn, Jeffrey L Goldberg
ABSTRACT
What pathways specify retinal ganglion cell (RGC) fate in the developing retina? Here we report on mechanisms by which a molecular pathway involving Sox4/Sox11 is required for RGC differentiation and for optic nerve formation in mice
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)