- Synthetic control of flowering in rice independent of the cultivation environment.
Synthetic control of flowering in rice independent of the cultivation environment.
Nature plants (2017-03-28)
Ryo Okada, Yasue Nemoto, Naokuni Endo-Higashi, Takeshi Izawa
PMID28346447
ABSTRACT
For genetically homogeneous crops, the timing of flowering is determined largely by the cultivation environment and is strongly associated with the yield and quality of the harvest