Merck
  • Separation and preconcentration of riboflavin from human plasma using polythionine coated magnetite/hydroxyapatite nanocomposite prior to analysis by surfactant-enhanced fluorimetry.
Separation and preconcentration of riboflavin from human plasma using polythionine coated magnetite/hydroxyapatite nanocomposite prior to analysis by surfactant-enhanced fluorimetry.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2017-05-12)
Leila Farzin, Mojtaba Shamsipur
ABSTRACT
The exploration of novel adsorption properties of conductive polymers based on hybridization with biocompatible nanomaterials receives an increasing interest. In this regard, hydroxyapatite (HA) bioceramic is of critical importance mainly owing to its facile synthesis, high surface area, economic and low toxicity in biological environments. In this work, we first prepared and characterized a magnetite/hydroxyapatite (Fe
Product Description
Supelco
Riboflavin (B2), analytical standard
Sigma-Aldrich
Pyridoxine, ≥98%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Lumichrome