Merck
  • Bioactive compound and antioxidant activity distribution in roller-milled and pearled fractions of conventional and pigmented wheat varieties.
Bioactive compound and antioxidant activity distribution in roller-milled and pearled fractions of conventional and pigmented wheat varieties.
Food chemistry (2017-05-23)
Debora Giordano, Monica Locatelli, Fabiano Travaglia, Matteo Bordiga, Amedeo Reyneri, Jean Daniel Coïsson, Massimo Blandino
ABSTRACT
In this study, the chemical composition of pigmented wheats (yellow, purple and blue types), and the distribution of the bioactive compounds in their roller-milled and pearled fractions, were compared with conventional wheats (red and white types). Roller-milling promoted the recovery of total dietary fiber, β-glucans, phenolic acids and anthocyanins in the bran fraction, which resulted also in a higher total antioxidant activity than the refined flour. Conversely, lutein resulted mainly concentrated in the refined flour. In the same way, the distribution pattern in the pearled fractions differ depending on the bioactive considered. The study highlights that a careful selection of the most appropriate fractionation process should be performed to produce flours rich in bioactive compounds. Roller-milling resulted useful for the production of refined flours rich in xanthophylls, with particular emphasis to the yellow-grained wheats. Contrarily, pearling could be more useful in the valorization of the health potential of anthocyanin-pigmented varieties.
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Sigma-Aldrich
Ellagic acid, ≥95% (HPLC), powder, from tree bark
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
Syringic acid, ≥95% (HPLC)
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)