Merck
  • Synthesis and toxicity assessment of 3-oxobutanamides against human lymphocytes and isolated mitochondria.
Synthesis and toxicity assessment of 3-oxobutanamides against human lymphocytes and isolated mitochondria.
Environmental toxicology and pharmacology (2017-03-16)
Nima Razzaghi-Asl, Enaytollah Seydi, Radin Alikhani, Saba Rezvani, Ramin Miri, Ahmad Salimi
ABSTRACT
To reduce costly late-phase compound scrubbing, there has been an increased focus on assessing compounds within in vitro assays that predict properties of human safety liabilities, before preclinical in vivo studies. The aim of our study was to answer the questions that whether the toxicity risk of a series of 3-oxobutanamide derivatives could be predicted by using of human lymphocytes and their isolated mitochondria. Using biochemical and flow cytometry assessments, we demonstrated that exposure of lymphocytes and isolated mitochondria to five 3-oxobutanamide derivatives (1-5) did not exhibit remarkable toxicity at low concentrations (50-500μM) but toxicity could be observed at high concentrations (1000 and 2000μM), particularly for N-(5-(4-bromophenyl)-3-isoxazolyl)-3-oxobutanamide (4) and N-(2-benzothiazolyl)-3-oxo butanamide (5). Compounds 4, 5 and partly N-(5-methyl-3-isoxazol yl)-3-oxo butanamide (1) also showed a marked cellular and mitochondrial toxicity while compound 5 displayed superior toxicity. Compound 5 induced cytotoxicity on human blood lymphocytes which was associated with the generation of intracellular reactive oxygen species (ROS), mitochondrial membrane potential (MMP) collapse, lysosomal membrane injury, lipid peroxidation and depletion of glutathione. Our results suggested that among assessed compounds, increased toxicity of compound 5 compared to other compounds could be likely attributed to the presence of bromine substituent in 5. Finally our findings proposed that using of antioxidants and mitochondrial/lysosomal protective agents could be beneficial in decreasing the toxicity of 5.
Sigma-Aldrich
ADP/ATP Ratio Assay Kit, sufficient for 100 tests (bioluminescent)