Merck
  • Exercise-induced circulating extracellular vesicles protect against cardiac ischemia-reperfusion injury.
Exercise-induced circulating extracellular vesicles protect against cardiac ischemia-reperfusion injury.
Basic research in cardiology (2017-05-24)
Yihua Bei, Tianzhao Xu, Dongchao Lv, Pujiao Yu, Jiahong Xu, Lin Che, Avash Das, John Tigges, Vassilios Toxavidis, Ionita Ghiran, Ravi Shah, Yongqin Li, Yuhui Zhang, Saumya Das, Junjie Xiao
ABSTRACT
Extracellular vesicles (EVs) serve an important function as mediators of intercellular communication. Exercise is protective for the heart, although the signaling mechanisms that mediate this cardioprotection have not been fully elucidated. Here using nano-flow cytometry, we found a rapid increase in plasma EVs in human subjects undergoing exercise stress testing. We subsequently identified that serum EVs were increased by ~1.85-fold in mice after 3-week swimming. Intramyocardial injection of equivalent quantities of EVs from exercised mice and non-exercised controls provided similar protective effects against acute ischemia/reperfusion (I/R) injury in mice. However, injection of exercise-induced EVs in a quantity equivalent to the increase seen with exercise (1.85 swim group) significantly enhanced the protective effect. Similarly, treatment with exercise-induced increased EVs provided additional anti-apoptotic effect in H
Sigma-Aldrich
Monoclonal Anti-α-Actinin (Sarcomeric) antibody produced in mouse, clone EA-53, ascites fluid
Sigma-Aldrich
PKH67 Green Fluorescent Cell Linker Kit for General Cell Membrane Labeling, Distributed for Phanos Technologies