- Synthesis and characterization of silver phosphate/calcium phosphate mixed particles capable of silver nanoparticle formation by photoreduction.
Synthesis and characterization of silver phosphate/calcium phosphate mixed particles capable of silver nanoparticle formation by photoreduction.
Materials science & engineering. C, Materials for biological applications (2017-05-10)
Livia C Natale, Yvette Alania, Marcela C Rodrigues, Alyne Simões, Douglas N de Souza, Erick de Lima, Victor E Arana-Chavez, Thiago L R Hewer, Rochelle Hiers, Fernando L Esteban-Florez, Giancarlo E S Brito, Sharukh Khajotia, Roberto R Braga
PMID28482551
ABSTRACT
Silver phosphate is a semi-conductor sensitive to UV-Vis radiation (<530nm). Exposure to radiation removes electrons from the oxygen valence shell, which are scavenged by silver cations (Ag
Product Description
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM