Merck
  • Testosterone and dihydrotestosterone levels in the transition zone correlate with prostate volume.
Testosterone and dihydrotestosterone levels in the transition zone correlate with prostate volume.
The Prostate (2017-06-09)
Tomislav Pejčić, Tomislav Tosti, Živoslav Tešić, Borivoj Milković, Dejan Dragičević, Milutin Kozomara, Milica Čekerevac, Zoran Džamić
ABSTRACT
There is still no consensus regarding intraprostatic androgen levels and the accumulation of androgens in the hyperplastic prostatic tissue. The current opinion is that intraprostatic dihydrotestosterone (DHT) concentrations are maintained but not elevated in benign prostatic hyperplasia (BPH), while there is no similar data concerning intraprostatic testosterone (T). Tissue T (tT) and tissue DHT (tDHT) concentration were determined in 93 patients scheduled for initial prostate biopsy. The criteria for biopsy were abnormal DRE and/or PSA > 4 ng/mL. Total prostate volume (TPV) was determined by transrectal ultrasound (TRUS). During TRUS- guided prostate biopsy, 10-12 samples were collected from the peripheral zone (PZ) and two additional samples were collected from the transition zone (TZ). The samples from the TZ were immediately frozen in liquid nitrogen at -70°C, and transported for tissue androgen determination, using liquid chromatography mass spectrometry (LC-MS). Pathological analysis revealed that prostate cancer (PCa) was present in 45 and absent in 48 patients. In the whole group, there were 42 men with small prostate (TPV < 30 mL) and 51 with enlarged prostate (TPV ≥ 31 mL). The overall average tT level was 0.79 ± 0.66 ng/g, while the average tDHT level was 10.27 ± 7.15 ng/g. There were no differences in tT and tDHT level in prostates with and without PCa. However, tT and tDHT levels were significantly higher in larger, than in smaller prostates (tT: 1.05 ± 0.75 and 0.46 ± 0.29 ng/g, and tDHT: 15.0 ± 6.09 and 4.51 ± 2.75 ng/g, respectively). There were strong correlations between tT and TPV (r = 0.71), and tDHT and TPV (r = 0.74). The present study confirmed that both T and DHT accumulated in the stroma of enlarged prostates; the degree of accumulation correlated with prostate volume.