Merck
  • Comparative secretomic analysis of lignocellulose degradation by Lentinula edodes grown on microcrystalline cellulose, lignosulfonate and glucose.
Comparative secretomic analysis of lignocellulose degradation by Lentinula edodes grown on microcrystalline cellulose, lignosulfonate and glucose.
Journal of proteomics (2017-05-10)
Yingli Cai, Yuhua Gong, Wei Liu, Yue Hu, Lianfu Chen, Lianlian Yan, Yan Zhou, Yinbing Bian
ABSTRACT
Lentinula edodes has the potential to degrade woody and nonwoody lignocellulosic biomass. However, the mechanism of lignocellulose degradation by L. edodes is unclear. The aim of this work is to explore the profiling of soluble secreted proteins involved in lignocellulose degradation in L. edodes. For that, we compared the secretomes of L. edodes grown on microcrystalline cellulose, cellulose with lignosulfonate and glucose. Based on nanoliquid chromatography coupled with tandem mass spectrometry of whole-protein hydrolysate, 230 proteins were identified. Label-free proteomic analysis showed that the most abundant carbohydrate-active enzymes involved in polysaccharide hydrolysis were endo-β-1,4-glucanase, α-galactosidase, polygalacturonase and glucoamylase in both cellulosic secretomes. In contrast, enzymes involved in lignin degradation were most abundant in glucose culture, with laccase 1 being the predominant protein (13.13%). When the cellulose and cellulose with lignosulfonate secretomes were compared, the abundance of cellulases and hemicellulases was higher in cellulose with lignosulfonate cultures, which was confirmed by enzyme activity assays. In addition, qRT-PCR analysis demonstrated that the expression levels of genes encoding cellulases and hemicellulases were significantly increased (by 32.2- to 1166.7-fold) when L. edodes was grown in cellulose with lignosulfonate medium. In this article, the secretomes of L. edodes grown on three different carbon sources were compared. The presented results revealed the profiling of extracellular enzymes involved in lignocellulose degradation, which is helpful to further explore the mechanism of biomass bioconversion by L. edodes.