Merck
  • A tumor-targeted polymer theranostics platform for positron emission tomography and fluorescence imaging.
A tumor-targeted polymer theranostics platform for positron emission tomography and fluorescence imaging.
Nanoscale (2017-07-22)
Eva Koziolová, Shreya Goel, Petr Chytil, Olga Janoušková, Todd E Barnhart, Weibo Cai, Tomáš Etrych
ABSTRACT
Here, we describe a novel polymer platform suitable for efficient diagnostics and potential theranostics based on
Sigma-Aldrich
2-Thiazoline-2-thiol, 98%
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)