Merck
  • Identification of novel targets for PGC-1alpha and histone deacetylase inhibitors in neuroblastoma cells.
Identification of novel targets for PGC-1alpha and histone deacetylase inhibitors in neuroblastoma cells.
Biochemical and biophysical research communications (2009-01-03)
Rita M Cowell, Pratik Talati, Kathryn R Blake, James H Meador-Woodruff, James W Russell
ABSTRACT
Recent evidence suggests that the transcriptional coactivator peroxisome proliferator activated receptor gamma coactivator 1alpha (PGC-1alpha) is involved in the pathology of Huntington's Disease (HD). While animals lacking PGC-1alpha express lower levels of genes involved in antioxidant defense and oxidative phosphorylation in the brain, little is known about other targets for PGC-1alpha in neuronal cells and whether there are ways to pharmacologically target PGC-1alpha in neurons. Here, PGC-1alpha overexpression in SH-SY5Y neuroblastoma cells upregulated expression of genes involved in mitochondrial function, glucose transport, fatty acid metabolism, and synaptic function. Overexpression also decreased vulnerability to hydrogen peroxide-induced cell death and caspase 3 activation. Treatment of cells with the histone deacetylase inhibitors (HDACi's) trichostatin A and valproic acid upregulated PGC-1alpha and glucose transporter 4 (GLUT4). These results suggest that PGC-1alpha regulates multiple pathways in neurons and that HDACi's may be good candidates to target PGC-1alpha and GLUT4 in HD and other neurological disorders.
Sigma-Aldrich
Monoclonal Anti-Synaptophysin antibody produced in mouse, clone SVP-38, ascites fluid
Sigma-Aldrich
Anti-Actin Antibody, clone C4, clone C4, Chemicon®, from mouse
Sigma-Aldrich
Anti-Green Fluorescent Protein Antibody, original GFP only, clone 264-449-2, clone 264-449-2, Chemicon®, from mouse
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture