Merck
Depleted upconversion luminescence in NaYF
Physical chemistry chemical physics : PCCP (2017-06-29)
Hongxin Zhang, Tianqing Jia, Long Chen, Yuchan Zhang, Shian Zhang, Donghai Feng, Zhenrong Sun, Jianrong Qiu
ABSTRACT
We report optical depletion of upconversion luminescence (UCL) in NaYF
Sigma-Aldrich
1-Octadecene, technical grade, 90%
Sigma-Aldrich
Yttrium(III) acetate hydrate, 99.9% metals basis
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Thulium(III) acetate hydrate, 99.9% trace metals basis
Sigma-Aldrich
Ytterbium(III) acetate hydrate, 99.95% trace metals basis