Merck
  • High-affinity cooperative Ca2+ binding by MICU1-MICU2 serves as an on-off switch for the uniporter.
High-affinity cooperative Ca2+ binding by MICU1-MICU2 serves as an on-off switch for the uniporter.
EMBO reports (2017-06-16)
Kimberli J Kamer, Zenon Grabarek, Vamsi K Mootha
ABSTRACT
The mitochondrial calcium uniporter is a Ca2+-activated Ca2+ channel that is essential for dynamic modulation of mitochondrial function in response to cellular Ca2+ signals. It is regulated by two paralogous EF-hand proteins-MICU1 and MICU2, but the mechanism is unknown. Here, we demonstrate that both MICU1 and MICU2 are stabilized by Ca2+ We reconstitute the MICU1-MICU2 heterodimer and demonstrate that it binds Ca2+ cooperatively with high affinity. We discover that both MICU1 and MICU2 exhibit affinity for the mitochondria-specific lipid cardiolipin. We determine the minimum Ca2+ concentration required for disinhibition of the uniporter in permeabilized cells and report a close match with the Ca2+-binding affinity of MICU1-MICU2. We conclude that cooperative, high-affinity interaction of the MICU1-MICU2 complex with Ca2+ serves as an on-off switch, leading to a tightly controlled channel, capable of responding directly to cytosolic Ca2+ signals.
Sigma-Aldrich
Anti-MICU1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-LRPPRC antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine, ≥97%