Merck
  • Oxygen Sensing with Perfluorocarbon-Loaded Ultraporous Mesostructured Silica Nanoparticles.
Oxygen Sensing with Perfluorocarbon-Loaded Ultraporous Mesostructured Silica Nanoparticles.
ACS nano (2017-05-16)
Amani L Lee, Clifford T Gee, Bradley P Weegman, Samuel A Einstein, Adam R Juelfs, Hattie L Ring, Katie R Hurley, Sam M Egger, Garrett Swindlehurst, Michael Garwood, William C K Pomerantz, Christy L Haynes
ABSTRACT
Oxygen homeostasis is important in the regulation of biological function. Disease progression can be monitored by measuring oxygen levels, thus producing information for the design of therapeutic treatments. Noninvasive measurements of tissue oxygenation require the development of tools with minimal adverse effects and facile detection of features of interest. Fluorine magnetic resonance imaging (
Sigma-Aldrich
N,N-Dimethylhexadecylamine, technical, ≥95% (GC)
Pricing and availability is not currently available.