Merck
Polyphenol patterns to trace sweet (
Journal of food science and technology (2017-07-26)
Gianluca Picariello, Pasquale Ferranti, Fausta De Cunzo, Ermelinda Sacco, Maria Grazia Volpe
ABSTRACT
The aim of this work was to assess whether the characteristic polyphenol traits of cherry biotypes persisted in thermally processed cherry products, such as jam. Thus, the RP-HPLC-diode array detector profiles of both colorless polyphenols and anthocyanins from three cherry varieties (two sweet and one tart cherry) were compared with those of low-sugar jam sourced from the same
Sigma-Aldrich
Quercitrin hydrate, ≥78%
Quercitrin, primary reference standard