Merck
  • NFAM1, an immunoreceptor tyrosine-based activation motif-bearing molecule that regulates B cell development and signaling.
NFAM1, an immunoreceptor tyrosine-based activation motif-bearing molecule that regulates B cell development and signaling.
Proceedings of the National Academy of Sciences of the United States of America (2004-05-15)
Makoto Ohtsuka, Hisashi Arase, Arata Takeuchi, Sho Yamasaki, Ritsuko Shiina, Tadahiro Suenaga, Daiju Sakurai, Tadashi Yokosuka, Noriko Arase, Makio Iwashima, Toshio Kitamura, Hideshige Moriya, Takashi Saito
ABSTRACT
A functional cDNA cloning system was developed by using a retrovirus library encoding CD8-chimeric proteins and a nuclear factor of activated T cells (NFAT)-GFP reporter cell line to identify molecules inducing NFAT activation. By using this strategy, NFAT activating molecule 1 (NFAM1) was cloned as an immunoreceptor tyrosine-based activation motif (ITAM)-bearing cell surface molecule belonging to the Ig superfamily and is predominantly expressed in spleen B and T cells. NFAM1 crosslinking induced ITAM phosphorylation, ZAP-70/Syk recruitment, NFAT activation, and cytokine production. In vivo overexpression of NFAM1 in bone marrow chimeras and transgenic mice induced severe impairment of early B cell development in an ITAM-dependent manner. In NFAM1-expressing B cells, B cell antigen receptor stimulation induced NFAM1 translocation to lipid raft, and NFAM1 co-crosslinking augmented B cell antigen receptor signaling. The results suggest that NFAM1 modulates B cell signaling through its ITAM, which regulates B cell development.
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)