Merck
  • Anti-glycation and anti-oxidative effects of a phenolic-enriched maple syrup extract and its protective effects on normal human colon cells.
Anti-glycation and anti-oxidative effects of a phenolic-enriched maple syrup extract and its protective effects on normal human colon cells.
Food & function (2017-01-24)
Weixi Liu, Zhengxi Wei, Hang Ma, Ang Cai, Yongqiang Liu, Jiadong Sun, Nicholas A DaSilva, Shelby L Johnson, Louis J Kirschenbaum, Bongsup P Cho, Joel A Dain, David C Rowley, Zahir A Shaikh, Navindra P Seeram
ABSTRACT
Oxidative stress and free radical generation accelerate the formation of advanced glycation endproducts (AGEs) which are linked to several chronic diseases. Published data suggest that phenolic-rich plant foods, show promise as natural anti-AGEs agents due to their anti-oxidation capacities. A phenolic-enriched maple syrup extract (MSX) has previously been reported to show anti-inflammatory and neuroprotective effects but its anti-AGE effects remain unknown. Therefore, herein, we investigated the anti-glycation and anti-oxidation effects of MSX using biochemical and biophysical methods. MSX (500 μg mL
Sigma-Aldrich
Aminoguanidine hydrochloride, ≥98%
Sigma-Aldrich
2′,7′-Dichlorodihydrofluorescein diacetate, ≥97%