Merck
  • Effects of Bundle Type and Substitution with Spent Laying Hen Surimi on Quality Characteristics of Imitation Crabsticks.
Effects of Bundle Type and Substitution with Spent Laying Hen Surimi on Quality Characteristics of Imitation Crabsticks.
Korean journal for food science of animal resources (2017-05-19)
Sang-Keun Jin, Jung-Seok Choi, Gap-Don Kim
ABSTRACT
The purpose of this study was to evaluate the effects of bundle type (BT) and substitution with spent laying hen (SH) surimi on quality characteristics of imitation crabsticks made from Alaska Pollack (AP) during 6 wk of cold storage. Diagonally bundled samples had poorer gel characteristics and more lipid oxidation when compared with straight bundled ones (
Product Description
Supelco
Supelco 37 Component FAME Mix, certified reference material, TraceCERT®, in dichloromethane (varied conc.), ampule of 1 mL