Merck
Interaction of VO2+ ion with human serum transferrin and albumin.
Journal of inorganic biochemistry (2009-02-10)
Daniele Sanna, Eugenio Garribba, Giovanni Micera
ABSTRACT
The complexation of VO(2+) ion with the high molecular mass components of the blood serum, human serum transferrin (hTf) and albumin (HSA), has been re-examined using EPR spectroscopy. In the case of transferrin, the results confirm those previously obtained, showing that VO(2+) ion occupies three different binding sites, A, B(1) and B(2), distinguishable in the X-band anisotropic spectrum recorded in D(2)O. With albumin the results show that a dinuclear complex (VO)(2)(d)HSA is formed in equimolar aqueous solutions or with an excess of protein; in the presence of an excess of VO(2+), the multinuclear complex (VO)(x)(m)HSA is the prevalent species, where x=5-6 indicates the equivalents of metal ion coordinated by HSA. The structure of the dinuclear species is discussed and the donor atoms involved in the metal coordination are proposed on the basis of the measured EPR parameters. Two different binding modes of albumin can be distinguished varying the pH, with only one species being present at the physiological value. The results show that the previously named "strong" site is not the N-terminal copper binding site, and some hypothesis on the metal coordination is discussed, with the (51)V A(z) values for the proposed donor sets obtained by DFT (density functional theory) calculations. Finally, preliminary results obtained in the ternary system VO(2+)/hTf/HSA are shown in order to determine the different binding strength of the two proteins. Due to the low VO(2+) concentration used, the recording of the EPR spectra through the repeated acquisition of the weak signals is essential to obtain a good signal to noise ratio in these systems.
Sigma-Aldrich
Albumin from human serum, lyophilized powder, ≥97% (agarose gel electrophoresis)