Merck
High-throughput fluorescence assay of cytochrome P450 3A4.
Nature protocols (2009-08-08)
Qian Cheng, Christal D Sohl, F Peter Guengerich
ABSTRACT
Cytochrome P450 mono-oxygenases (P450s) are the principal enzymes involved in the oxidative metabolism of drugs and other xenobiotics. In this protocol, we describe a fluorescence-based, high-throughput assay for measuring the activity of P450 3A4, one of the key enzymes involved in drug metabolism. The assay involves the oxidative debenzylation of a substituted coumarin, yielding an increase in fluorescence on reaction. The entire procedure can be accomplished in 1 h or less.
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type XV, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline