Merck
  • The Arabidopsis DJ-1a protein confers stress protection through cytosolic SOD activation.
The Arabidopsis DJ-1a protein confers stress protection through cytosolic SOD activation.
Journal of cell science (2010-04-22)
Xiang Ming Xu, Hong Lin, Jodi Maple, Benny Björkblom, Guido Alves, Jan Petter Larsen, Simon Geir Møller
ABSTRACT
Mutations in the DJ-1 gene (also known as PARK7) cause inherited Parkinson's disease, which is characterized by neuronal death. Although DJ-1 is thought to be an antioxidant protein, the underlying mechanism by which loss of DJ-1 function contributes to cell death is unclear. Human DJ-1 and its Arabidopsis thaliana homologue, AtDJ-1a, are evolutionarily conserved proteins, indicating a universal function. To gain further knowledge of the molecular features associated with DJ-1 dysfunction, we have characterized AtDJ-1a. We show that AtDJ-1a levels are responsive to stress treatment and that AtDJ-1a loss of function results in accelerated cell death in aging plants. By contrast, transgenic plants with elevated AtDJ-1a levels have increased protection against environmental stress conditions, such as strong light, H(2)O(2), methyl viologen and copper sulfate. We further identify superoxide dismutase 1 (SOD1) and glutathione peroxidase 2 (GPX2) as interaction partners of both AtDJ-1a and human DJ-1, and show that this interaction results in AtDJ-1a- and DJ-1-mediated cytosolic SOD1 activation in a copper-dependent fashion. Our data have highlighted a conserved molecular mechanism for DJ-1 and revealed a new protein player in the oxidative stress response of plants.
Sigma-Aldrich
BrdU (5-Bromo-2′-deoxyuridine), Used for the study of DNA synthesis where it is incorporated into DNA in place of thymidine, BrdU can be used in conjunction with anti-BrdU for immunocytochemical analysis of cell proliferation.
Sigma-Aldrich
SOD Assay Kit, sufficient for 500 tests