- Selective removal of alkynes from diene mixtures using ether-functionalized Cu(i)-containing ionic liquids as extractants.
Selective removal of alkynes from diene mixtures using ether-functionalized Cu(i)-containing ionic liquids as extractants.
Physical chemistry chemical physics : PCCP (2017-01-04)
Young Jin Kim, Hyun Ji Lee, Yohan Ahn, Shuyao Zhang, Je Seung Lee, Minserk Cheong, Hoon Sik Kim
PMID28045142
ABSTRACT
Cu(i)-Containing room temperature ionic liquids (Cu-EnA), prepared from CuCl and ether-functionalized ionic liquids (ILs) bearing a methanesulfonate anion (EnA, n = 1, 2 or 3), were thermally stable and highly effective for the removal of alkynes such as isopropenylacetylene (IPA) and 2-butyne (2-BT) contained in dienes like isoprene (2-methyl-1,3-butadiene). Cu-EnA were found to reversibly and selectively interact with IPA and 2-BT, thereby enabling the regeneration of Cu-EnA. Fast atom bombardment (FAB)-mass spectral and computational results imply that EnA consists of an ether-functionalized imidazolium cation and a methanesulfonate-coordinated Cu(i) anion such as [CuCl(CH