Skip to Content
CZ
EN
Products
Applications
Services
Resources
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Lab Automation
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Documents
Learning Center
New
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Home
Search Results
Concurrent activation of β
Concurrent activation of β
Cellular signalling (2017-05-13)
Artur Wnorowski, Justyna Such, Rajib K Paul, Robert P Wersto, Fred E Indig, Krzysztof Jozwiak, Michel Bernier, Irving W Wainer
PMID
28495590
ABSTRACT
Activation of β
Product Description
S4069
Sigma-Aldrich
SL327, ≥98% (HPLC), Mixture of E & Z isomers, solid
Pricing
A6226
Sigma-Aldrich
AM251, >98% (HPLC), solid
Pricing