Merck
Synthesis and pK
Chirality (2017-06-27)
Tamás Németh, Gergő Dargó, József Levente Petró, Zsófia Petrik, Sándor Lévai, Balázs Krámos, Zoltán Béni, József Nagy, György Tibor Balogh, Péter Huszthy, Tünde Tóth
ABSTRACT
New enantiopure dimethyl-substituted acridino-18-crown-6 and acridino-21-crown-7 ethers containing a carboxyl group at position 9 of the acridine ring [(S,S)-8, (S,S)-9, (R,R)-10] were synthesized. The pK
Sigma-Aldrich
Thymine 1-β-D-arabinofuranoside, ≥99%