Merck
  • Prediction of Relapse After Therapy Withdrawal in Women with Endometrial Hyperplasia: A Long-term Follow-up Study.
Prediction of Relapse After Therapy Withdrawal in Women with Endometrial Hyperplasia: A Long-term Follow-up Study.
Anticancer research (2017-05-10)
Elise Thoresen Sletten, Marit Arnes, Lena Myreng Lysa, Bjorn Tg Moe, Bjorn Straume, Anne Orbo
ABSTRACT
To investigate whether risk of relapse of endometrial hyperplasia persists many years after successful primary therapy and whether clinical or biological markers observed at primary diagnosis may predict relapse. A series of 57 women with endometrial hyperplasia received levonorgestrel-impregnated intrauterine system or oral progestin for three months during 1998-2000. Index biopsies were classified according to WHO1994 and D-score systems, and immunohistochemical staining for estrogen receptor α (ERα), estrogen receptor β (ERβ), progesterone receptor A (PRA), progesterone receptor B (PRB), B-cell lymphoma 2/apoptosis regulator (BCL2), BCL2-associated X protein/apoptosis regulator (BAX), paired box 2 (PAX2), and phosphatase and tensin homolog (PTEN) reported as H-scores. Over a follow-up of 157.8 months, 23% (10/43) of patients experienced relapse. No correlation with age, body mass index, parity, WHO94 classification, or D-score was found. Only PRA (p=0.004) and PRB (p=0.038) showed certain correlation with relapse. Endometrial hyperplasia recurs many years after successful progestin therapy. Increased expression of PRB and reduced expression of PRA significantly correlated with relapse. Our results support the importance of continuous endometrial protection and the need for new clinical surveillance guidelines.
Product Description
Supelco
ORBO 24 2-HMP on Amberlite XAD®-2 (20/40), 150/75 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 25 ea
Supelco
ORBO 43 Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Supelco
ORBO 65P (OVS) Amberlite XAD®-4 specially cleaned, 160/80 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS), pkg of 10 ea
Supelco
ORBO 92 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group Vinyl acetate
Supelco
ORBO 80 Isocyanate Filter, with cassettes: no, filter O.D. 37 mm, pkg of 25 ea
Supelco
ORBO 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 402 Tenax® TA specially treated (35/60), 100/50 mg, W,W,W separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 53 Activated Silica Gel (20/40) specially cleaned, 400/200 mg with Glass Fiber Filters, GFF,F,F separators, O.D. × L 7 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 502 Activated Silica Gel (20/45) 100/50 mg, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 554 H2SO4 on Silica Gel (20/40) 150/75 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 80 Coated Filter, with cassettes unassembled
Supelco
ORBO 609 Amberlite XAD®-2 (20/50) 400/200 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
Supelco
ORBO 23 2-HMP on Amberlite XAD®-2 (20/40), 120/60 mg, W,W,W separators, O.D. × L 6 mm × 85 mm, pkg of 25 ea
Supelco
ORBO 49P (OVS) Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 270/140 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube, pkg of 10 ea