Merck
  • Differential Off-line LC-NMR (DOLC-NMR) Metabolomics To Monitor Tyrosine-Induced Metabolome Alterations in Saccharomyces cerevisiae.
Differential Off-line LC-NMR (DOLC-NMR) Metabolomics To Monitor Tyrosine-Induced Metabolome Alterations in Saccharomyces cerevisiae.
Journal of agricultural and food chemistry (2017-04-07)
Richard Hammerl, Oliver Frank, Thomas Hofmann
ABSTRACT
A novel differential off-line LC-NMR approach (DOLC-NMR) was developed to capture and quantify nutrient-induced metabolome alterations in Saccharomyces cerevisiae. Off-line coupling of HPLC separation and
Sigma-Aldrich
4-Hydroxyphenethyl bromide, 96%
Pricing and availability is not currently available.