Merck
  • In vitro investigation of anticancer and ACE-inhibiting activity, α-amylase and α-glucosidase inhibition, and antioxidant activity of camel milk fermented with camel milk probiotic: A comparative study with fermented bovine milk.
In vitro investigation of anticancer and ACE-inhibiting activity, α-amylase and α-glucosidase inhibition, and antioxidant activity of camel milk fermented with camel milk probiotic: A comparative study with fermented bovine milk.
Food chemistry (2017-09-07)
Mutamed Ayyash, Amna K Al-Nuaimi, Suheir Al-Mahadin, Shao-Quan Liu
ABSTRACT
This study aimed to investigate in vitro the health-promoting benefits (anticancer activity, α-amylase and α-glucosidase inhibition, angiotensin-converting-enzyme (ACE)-inhibition, antioxidant and proteolytic activity) of camel milk fermented with indigenous probiotic strains of Lactobacillus spp., compared with fermented bovine milk. The three camel milk probiotic strains Lb. reuteri-KX881777, Lb. plantarum-KX881772, Lb. plantarum-KX881779 and a control strain Lb. plantarum DSM2468 were employed to ferment camel and bovine milks separately. The proteolytic and antioxidant activity of water soluble extracts (WSEs) from all fermented camel milks were higher than those of fermented bovine milk. α-Amylase inhibition of WSEs were >34% in both milk types fermented with all strains during storage periods, except the WSE of camel milk fermented by Lp.K772. The highest ACE-inhibition of the WSE from camel milk fermented by Lr.K777 was >80%. The proliferations of Caco-2, MCF-7 and HELA cells were more inhibited when treated with the WSE of fermented camel milk.
Sigma-Aldrich
N-Hexanoyl-L-homoserine lactone, ≥96% (HPLC)