Merck
  • Coculture of mesenchymal stem cells and endothelial cells enhances host tissue integration and epidermis maturation through AKT activation in gelatin methacryloyl hydrogel-based skin model.
Coculture of mesenchymal stem cells and endothelial cells enhances host tissue integration and epidermis maturation through AKT activation in gelatin methacryloyl hydrogel-based skin model.
Acta biomaterialia (2017-07-08)
Xiaofei Zhang, Jun Li, Pengxiang Ye, Guifang Gao, Karen Hubbell, Xiaofeng Cui
ABSTRACT
A major challenge for clinical use of skin substitutes is insufficient host tissue integration leading to loosening and partial necrosis of the implant. In this present study, a three-dimensional (3D) coculture system constructed using human umbilical cord mesenchymal stem cells (uc-MSCs) and umbilical vein endothelial cells (HUVECs) encapsulated in gelatin methacryloyl (GelMA) hydrogels was evaluated to determine the outcomes of cell-cell interactions in vitro and in vivo. The results revealed that GelMA hydrogels displayed minor cytotoxicity on both cell types. An uc-MSC:HUVEC ratio of 50:50 demonstrated the highest cell proliferation and expression of angiogenic markers. The supplement of basic fibroblast growth factors (bFGF) in coculture system further induced cell proliferation and gene expression in vitro. In vivo transplantation of this cocultured constructs efficiently enhanced the implant and host tissue integration. Additionally, the proliferation of keratinocytes was well maintained on GelMA hydrogels and the gene expression related to cell proliferation and differentiation was significantly increased in coculture system comparing to monoculture. Mechanistically, AKT signaling pathways were activated in cocultures. Our findings suggest that coculturing MSC and EC in GelMA hydrogels could be a promising approach to substantially improve the integration of exogenous skin substitutes and host tissues. In this study, the co-culture of uc-MSCs and HUVECs in photocrosslinkable GelMA hydrogels significantly enhanced host tissue integration. Cell proliferation, ECM deposition and angiogenic genes expression were all substantially improved in vitro and the excellent host tissue integration into the implanted tissue was observed in vivo. When served as a dermal layer, the scaffold with co-cultured cells enhanced the proliferation and differentiation of keratinocytes. AKT signaling was proved to be involved in the regulation of cell survival and fate determination. This work demonstrated the importance of 3D cell co-culture to facilitate host tissue integration that can be a promising approach for long-term survival of skin substitutes.
Sigma-Aldrich
Endothelial cell growth supplement from bovine neural tissue, ECGS, suitable for cell culture
Sigma-Aldrich
Nutrient Mixture F-12 Ham, Kaighn′s Modification, with L-glutamine, without sodium bicarbonate, powder, suitable for cell culture