Merck
  • Phenotype-driven precision oncology as a guide for clinical decisions one patient at a time.
Phenotype-driven precision oncology as a guide for clinical decisions one patient at a time.
Nature communications (2017-09-07)
Shumei Chia, Joo-Leng Low, Xiaoqian Zhang, Xue-Lin Kwang, Fui-Teen Chong, Ankur Sharma, Denis Bertrand, Shen Yon Toh, Hui-Sun Leong, Matan T Thangavelu, Jacqueline S G Hwang, Kok-Hing Lim, Thakshayeni Skanthakumar, Hiang-Khoon Tan, Yan Su, Siang Hui Choo, Hannes Hentze, Iain B H Tan, Alexander Lezhava, Patrick Tan, Daniel S W Tan, Giridharan Periyasamy, Judice L Y Koh, N Gopalakrishna Iyer, Ramanuj DasGupta
ABSTRACT
Genomics-driven cancer therapeutics has gained prominence in personalized cancer treatment. However, its utility in indications lacking biomarker-driven treatment strategies remains limited. Here we present a "phenotype-driven precision-oncology" approach, based on the notion that biological response to perturbations, chemical or genetic, in ex vivo patient-individualized models can serve as predictive biomarkers for therapeutic response in the clinic. We generated a library of "screenable" patient-derived primary cultures (PDCs) for head and neck squamous cell carcinomas that reproducibly predicted treatment response in matched patient-derived-xenograft models. Importantly, PDCs could guide clinical practice and predict tumour progression in two n = 1 co-clinical trials. Comprehensive "-omics" interrogation of PDCs derived from one of these models revealed YAP1 as a putative biomarker for treatment response and survival in ~24% of oral squamous cell carcinoma. We envision that scaling of the proposed PDC approach could uncover biomarkers for therapeutic stratification and guide real-time therapeutic decisions in the future.Treatment response in patient-derived models may serve as a biomarker for response in the clinic. Here, the authors use paired patient-derived mouse xenografts and patient-derived primary culture models from head and neck squamous cell carcinomas, including metastasis, as models for high-throughput screening of anti-cancer drugs.
Sigma-Aldrich
Pyridinium dichromate, 98%
Sigma-Aldrich
(2-Hydroxypropyl)-β-cyclodextrin, average Mw ~1,460
Sigma-Aldrich
TWEEN® 80, suitable for cell culture, suitable for insect cell culture, viscous liquid
Sigma-Aldrich
MISSION® esiRNA, targeting human HIST2H3C, HIST2H3A
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Millipore
Protease Inhibitor Cocktail Set III, EDTA-Free, Protease inhibitor cocktail III, EDTA-free for inhibiting aspartic, cysteine, and serine proteases as well as aminopeptidases in mammalian cells and tissues.