- High photocatalytic activity of hierarchical SiO
High photocatalytic activity of hierarchical SiO
Journal of hazardous materials (2017-07-19)
Ying Zhang, Juanrong Chen, Li Hua, Songjun Li, Xuanxuan Zhang, Weichen Sheng, Shunsheng Cao
PMID28719847
ABSTRACT
Ongoing research activities are targeted to explore high photocatalytic activity of TiO