Merck
  • Hypoxia-responsive ionizable liposome delivery siRNA for glioma therapy.
Hypoxia-responsive ionizable liposome delivery siRNA for glioma therapy.
International journal of nanomedicine (2017-02-23)
Hong-Mei Liu, Ya-Fei Zhang, Yan-Dong Xie, Yi-Fan Cai, Bai-Yang Li, Wen Li, Ling-Yu Zeng, Yu-Ling Li, Ru-Tong Yu
ABSTRACT
Here, we report the hypoxia-responsive ionizable liposomes to deliver small interference RNA (siRNA) anticancer drugs, which can selectively enhance cellular uptake of the siRNA under hypoxic and low-pH conditions to cure glioma. For this purpose, malate dehydrogenase lipid molecules were synthesized, which contain nitroimidazole groups that impart hypoxia sensitivity and specificity as hydrophobic tails, and tertiary amines as hydrophilic head groups. These malate dehydrogenase molecules, together with DSPE-PEG2000 and cholesterol, were self-assembled into O'
Sigma-Aldrich
MISSION® esiRNA, targeting human PLK1
Sigma-Aldrich
2-(Dimethylamino)pyridine, 97%