Merck
  • Improved respirable fraction of budesonide powder for dry powder inhaler formulations produced by advanced supercritical CO
Improved respirable fraction of budesonide powder for dry powder inhaler formulations produced by advanced supercritical CO
International journal of pharmaceutics (2017-06-07)
Yuta Miyazaki, Naoki Aruga, Kazunori Kadota, Yuichi Tozuka, Hirofumi Takeuchi
ABSTRACT
A budesonide (BDS) suspension was obtained via advanced supercritical carbon dioxide (scCO
Sigma-Aldrich
Sorbitan monopalmitate
Pricing and availability is not currently available.