Merck
  • Formation and purification of tailored liposomes for drug delivery using a module-based micro continuous-flow system.
Formation and purification of tailored liposomes for drug delivery using a module-based micro continuous-flow system.
Scientific reports (2017-09-25)
Nikolay Dimov, Elisabeth Kastner, Maryam Hussain, Yvonne Perrie, Nicolas Szita
ABSTRACT
Liposomes are lipid based bilayer vesicles that can encapsulate, deliver and release low-soluble drugs and small molecules to a specific target site in the body. They are currently exploited in several nanomedicine formulations. However, their development and application is still limited by expensive and time-consuming process development and production methods. Therefore, to exploit these systems more effectively and support the rapid translation of new liposomal nanomedicines from bench to bedside, new cost-effective and scalable production methods are needed. We present a continuous process flow system for the preparation, modification and purification of liposomes which offers lab-on-chip scale production. The system was evaluated for a range of small vesicles (below 300 nm) varying in lipid composition, size and charge; it offers effective and rapid nanomedicine purification with high lipid recovery (> 98%) combined with effective removal of non-entrapped drug (propofol >95% reduction of non-entrapped drug present) or protein (ovalbumin >90% reduction of OVA present) and organic solvent (ethanol >95% reduction) in less than 4 minutes. The key advantages of using this bench-top, rapid, process development tool are the flexible operating conditions, interchangeable membranes and scalable high-throughput yields, thereby offering simultaneous manufacturing and purification of nanoparticles with tailored surface attributes.
Sigma-Aldrich
N,N-Dimethyldodecylamine, 97%