Merck
  • The extent of intrauterine growth restriction determines the severity of cerebral injury and neurobehavioural deficits in rodents.
The extent of intrauterine growth restriction determines the severity of cerebral injury and neurobehavioural deficits in rodents.
PloS one (2017-09-22)
Crystal A Ruff, Stuart D Faulkner, Prakasham Rumajogee, Stephanie Beldick, Warren Foltz, Jennifer Corrigan, Alfred Basilious, Shangjun Jiang, Shanojan Thiyagalingam, Jerome Y Yager, Michael G Fehlings
ABSTRACT
Cerebral Palsy (CP) is the most common physical pediatric neurodevelopmental disorder and spastic diplegic injury is its most frequent subtype. CP results in substantial neuromotor and cognitive impairments that have significant socioeconomic impact. Despite this, its underlying pathophysiological mechanisms and etiology remain incompletely understood. Furthermore, there is a need for clinically relevant injury models, which a) reflect the heterogeneity of the condition and b) can be used to evaluate new translational therapies. To address these key knowledge gaps, we characterized a chronic placental insufficiency (PI) model, using bilateral uterine artery ligation (BUAL) of dams. This injury model results in intrauterine growth restriction (IUGR) in pups, and animals recapitulate the human phenotype both in terms of neurobehavioural and anatomical deficits. Effects of BUAL were studied using luxol fast blue (LFB)/hematoxylin & eosin (H&E) staining, immunohistochemistry, quantitative Magnetic Resonance Imaging (MRI), and Catwalk neurobehavioural tests. Neuroanatomical analysis revealed regional ventricular enlargement and corpus callosum thinning in IUGR animals, which was correlated with the extent of growth restriction. Olig2 staining revealed reductions in oligodendrocyte density in white and grey matter structures, including the corpus callosum, optic chiasm, and nucleus accumbens. The caudate nucleus, along with other brain structures such as the optic chiasm, internal capsule, septofimbrial and lateral septal nuclei, exhibited reduced size in animals with IUGR. The size of the pretectal nucleus was reduced only in moderately injured animals. MAG/NF200 staining demonstrated reduced myelination and axonal counts in the corpus callosum of IUGR animals. NeuN staining revealed changes in neuronal density in the hippocampus and in the thickness of hippocampal CA2 and CA3 regions. Diffusion weighted imaging (DWI) revealed regional white and grey matter changes at 3 weeks of age. Furthermore, neurobehavioural testing demonstrated neuromotor impairments in animals with IUGR in paw intensities, swing speed, relative print positions, and phase dispersions. We have characterized a rodent model of IUGR and have demonstrated that the neuroanatomical and neurobehavioural deficits mirror the severity of the IUGR injury. This model has the potential to be applied to examine the pathobiology of and potential therapeutic strategies for IUGR-related brain injury. Thus, this work has potential translational relevance for the study of CP.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Myelin Associated Glycoprotein Antibody, clone 513, clone 513, Chemicon®, from mouse
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Neurofilament 200 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution