Merck
  • BNIP3L/NIX-mediated mitophagy protects against ischemic brain injury independent of PARK2.
BNIP3L/NIX-mediated mitophagy protects against ischemic brain injury independent of PARK2.
Autophagy (2017-08-19)
Yang Yuan, Yanrong Zheng, Xiangnan Zhang, Ying Chen, Xiaoli Wu, Jiaying Wu, Zhe Shen, Lei Jiang, Lu Wang, Wei Yang, Jianhong Luo, Zhenghong Qin, Weiwei Hu, Zhong Chen
ABSTRACT
Cerebral ischemia induces massive mitochondrial damage. These damaged mitochondria are cleared, thus attenuating brain injury, by mitophagy. Here, we identified the involvement of BNIP3L/NIX in cerebral ischemia-reperfusion (I-R)-induced mitophagy. Bnip3l knockout (bnip3l-/-) impaired mitophagy and aggravated cerebral I-R injury in mice, which can be rescued by BNIP3L overexpression. The rescuing effects of BNIP3L overexpression can be observed in park2-/- mice, which showed mitophagy deficiency after I-R. Interestingly, bnip3l and park2 double-knockout mice showed a synergistic mitophagy deficiency with I-R treatment, which further highlighted the roles of BNIP3L-mediated mitophagy as being independent from PARK2. Further experiments indicated that phosphorylation of BNIP3L serine 81 is critical for BNIP3L-mediated mitophagy. Nonphosphorylatable mutant BNIP3LS81A failed to counteract both mitophagy impairment and neuroprotective effects in bnip3l-/- mice. Our findings offer insights into mitochondrial quality control in ischemic stroke and bring forth the concept that BNIP3L could be a potential therapeutic target for ischemic stroke, beyond its accepted role in reticulocyte maturation.
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Millipore
FLAG® Immunoprecipitation Kit
Sigma-Aldrich
Poly-L-lysine hydrobromide, mol wt 150,000-300,000